Merck
  • Preparative purification of a high-mannose type N-glycan from soy bean agglutinin by hydrazinolysis and tyrosinamide derivatization.
Preparative purification of a high-mannose type N-glycan from soy bean agglutinin by hydrazinolysis and tyrosinamide derivatization.
Analytical biochemistry (1999-01-12)
D L Evers, R L Hung, V H Thomas, K G Rice
ZUSAMMENFASSUNG
The N-linked oligosaccharide from soy bean agglutinin (Man9) was isolated on a preparative scale following derivatization with Boc-tyrosine. The procedure utilized preparative hydrazinolysis to release the oligosaccharide and yielded multi-micromol quantities of Boc-tyrosine-Man9 which was characterized by 1H NMR and ES-MS.