Merck
[The treatment of intra-abdominal abscess].
Klinichna khirurhiia (1999-02-16)
O A Beliaeva
ZUSAMMENFASSUNG
The experience of treatment of 18 patients with intraabdominal abscess using the regional laparostomy conduction and introducing nitazol, immobilized on polymethylsiloxane, in the abscess cavity, was summarized.