Merck
  • Noncanonical MMS2-encoded ubiquitin-conjugating enzyme functions in assembly of novel polyubiquitin chains for DNA repair.
Noncanonical MMS2-encoded ubiquitin-conjugating enzyme functions in assembly of novel polyubiquitin chains for DNA repair.
Cell (1999-03-25)
R M Hofmann, C M Pickart
ZUSAMMENFASSUNG
Ubiquitin-conjugating enzyme variant (UEV) proteins resemble ubiquitin-conjugating enzymes (E2s) but lack the defining E2 active-site residue. The MMS2-encoded UEV protein has been genetically implicated in error-free postreplicative DNA repair in Saccharomyces cerevisiae. We show that Mms2p forms a specific heteromeric complex with the UBC13-encoded E2 and is required for the Ubc13p-dependent assembly of polyubiquitin chains linked through lysine 63. A ubc13 yeast strain is UV sensitive, and single, double, and triple mutants of the UBC13, MMS2, and ubiquitin (ubiK63R) genes display a comparable phenotype. These findings support a model in which an Mms2p/Ubc13p complex assembles novel polyubiquitin chains for signaling in DNA repair, and they suggest that UEV proteins may act to increase diversity and selectivity in ubiquitin conjugation.