- Novel proton acceptor immonium-type coupling reagents: application in solution and solid-phase peptide synthesis.
Novel proton acceptor immonium-type coupling reagents: application in solution and solid-phase peptide synthesis.
Organic letters (2007-09-29)
Ayman El-Faham, Fernando Albericio
PMID17900195
ZUSAMMENFASSUNG
A novel proton acceptor coupling reagent shows superiority to those described previously. The oxygen in the carbocation moiety confers more solubility to the reagent. Furthermore, it enhances coupling yields and decreases racemization, allowing the use of 1 equiv of base.