Merck
RecQL4 helicase amplification is involved in human breast tumorigenesis.
PloS one (2013-07-31)
Hongbo Fang, Linghu Nie, Zhenfen Chi, Jing Liu, Dan Guo, Xuemei Lu, Tom K Hei, Adayabalam S Balajee, Yongliang Zhao
ZUSAMMENFASSUNG
Breast cancer occur both in hereditary and sporadic forms, and the later one comprises an overwhelming majority of breast cancer cases among women. Numerical and structural alterations involving chromosome 8, with loss of short arm (8p) and gain of long arm (8q), are frequently observed in breast cancer cells and tissues. In this study, we show that most of the human breast tumor cell lines examined display an over representation of 8q24, a chromosomal locus RecQL4 is regionally mapped to, and consequently, a markedly elevated level of RecQL4 expression. An increased RecQL4 mRNA level was also observed in a majority of clinical breast tumor samples (38/43) examined. shRNA-mediated RecQL4 suppression in MDA-MB453 breast cancer cells not only significantly inhibit the in vitro clonogenic survival and in vivo tumorigenicity. Further studies demonstrate that RecQL4 physically interacts with a major survival factor-survivin and its protein level affects survivin expression. Although loss of RecQL4 function due to gene mutations causally linked to occurrence of human RTS with features of premature aging and cancer predisposition, our studies provide the evidence that overexpression of RecQL4 due to gene amplification play a critical role in human breast tumor progression.