Merck
  • Interleukin-23 and interleukin-27 exert quite different antitumor and vaccine effects on poorly immunogenic melanoma.
Interleukin-23 and interleukin-27 exert quite different antitumor and vaccine effects on poorly immunogenic melanoma.
Cancer research (2006-06-17)
Shuntaro Oniki, Hiroshi Nagai, Tatsuya Horikawa, Junya Furukawa, Maria Laura Belladonna, Takayuki Yoshimoto, Isao Hara, Chikako Nishigori
ZUSAMMENFASSUNG
Recent studies revealed that two novel interleukin (IL)-12-related cytokines, IL-23 and IL-27, have potent antitumor activities. However, the antitumor effects were mainly evaluated in relatively highly immunogenic tumors and have not been fully evaluated against nonimmunogenic or poorly immunogenic tumors. In this study, we investigated the antitumor efficacies of IL-23 and IL-27 on poorly immunogenic B16F10 melanoma and found that the antitumor responses mediated by IL-23 and IL-27 were clearly different. In syngeneic mice, mouse single-chain (sc) IL-23-transfected B16F10 (B16/IL-23) tumors exhibited almost the same growth curve as B16F10 parental tumor about until day 20 after tumor injection and then showed growth inhibition or even regression. In contrast, scIL-27-transfected B16F10 (B16/IL-27) tumors exhibited significant retardation of tumor growth from the early stage. In vivo depletion assay revealed that the antitumor effect of B16/IL-23 was mainly mediated by CD8+ T cells and IFN-gamma whereas that of B16/IL-27 mainly involved natural killer cells and was independent of IFN-gamma. We also found that antitumor effects of B16/IL-23 and B16/IL-27 were synergistically enhanced by treatment with IL-18 and IL-12, respectively. Furthermore, B16/IL-23-vaccinated mice developed protective immunity against parental B16F10 tumors but B16/IL-27-vaccinated mice did not. When combined with prior in vivo depletion of CD25+ T cells, 80% of B16/IL-23-vaccinated mice completely rejected subsequent tumor challenge. Finally, we showed that the systemic administration of neither IL-23 nor IL-27 induced such intense toxicity as IL-12. Our data support that IL-23 and IL-27 might play a role in future cytokine-based immunotherapy against poorly immunogenic tumors.