Merck
Tetrastatin, the NC1 domain of the α4(IV) collagen chain: a novel potent anti-tumor matrikine.
PloS one (2012-04-28)
Sylvie Brassart-Pasco, Karine Sénéchal, Jessica Thevenard, Laurent Ramont, Jérome Devy, Ludivine Di Stefano, Aurélie Dupont-Deshorgue, Stéphane Brézillon, Jezabel Feru, Jean-François Jazeron, Marie-Danièle Diebold, Sylvie Ricard-Blum, François-Xavier Maquart, Jean Claude Monboisse
ZUSAMMENFASSUNG
NC1 domains from α1, α2, α3 and α6(IV) collagen chains were shown to exert anti-tumor or anti-angiogenic activities, whereas the NC1 domain of the α4(IV) chain did not show such activities so far. We demonstrate in the present paper that the NC1 α4(IV) domain exerts a potent anti-tumor activity both in vitro and in an experimental human melanoma model in vivo. The overexpression of NC1 α4(IV) in human UACC-903 melanoma cells strongly inhibited their in vitro proliferative (-38%) and invasive (-52%) properties. MT1-MMP activation was largely decreased and its cellular distribution was modified, resulting in a loss of expression at the migration front associated with a loss of migratory phenotype. In an in vivo xenograft model in athymic nude mice, the subcutaneous injection of NC1 α4(IV)-overexpressing melanoma cells induced significantly smaller tumors (-80% tumor volume) than the Mock cells, due to a strong inhibition of tumor growth. Exogenously added recombinant human NC1 α4(IV) reproduced the inhibitory effects of NC1 α4(IV) overexpression in UACC-903 cells but not in dermal fibroblasts. An anti-αvβ3 integrin blocking antibody inhibited cell adhesion on recombinant human NC1 α4(IV) substratum. The involvement of αvβ3 integrin in mediating NC1 α4(IV) effect was confirmed by surface plasmon resonance (SPR) binding assays showing that recombinant human NC1 α4(IV) binds to αvβ3 integrin (K(D) = 148 ± 9.54 nM). Collectively, our results demonstrate that the NC1 α4(IV) domain, named tetrastatin, is a new endogenous anti-tumor matrikine.