Merck
1H-NMR spectroscopy of body fluids: inborn errors of purine and pyrimidine metabolism.
Clinical chemistry (1999-04-02)
R A Wevers, U F Engelke, S H Moolenaar, C Bräutigam, J G de Jong, R Duran, R A de Abreu, A H van Gennip
ZUSAMMENFASSUNG
The diagnosis of inborn errors of purine and pyrimidine metabolism is often difficult. We examined the potential of 1H-NMR as a tool in evaluation of patients with these disorders. We performed 1H-NMR spectroscopy on 500 and 600 MHz instruments with a standardized sample volume of 500 microL. We studied body fluids from 25 patients with nine inborn errors of purine and pyrimidine metabolism. Characteristic abnormalities could be demonstrated in the 1H-NMR spectra of urine samples of all patients with diseases in the pyrimidine metabolism. In most urine samples from patients with defects in the purine metabolism, the 1H-NMR spectrum pointed to the specific diagnosis in a straightforward manner. The only exception was a urine from a case of adenine phosphoribosyl transferase deficiency in which the accumulating metabolite, 2,8-dihydroxyadenine, was not seen under the operating conditions used. Similarly, uric acid was not measured. We provide the 1H-NMR spectral characteristics of many intermediates in purine and pyrimidine metabolism that may be relevant for future studies in this field. The overview of metabolism that is provided by 1H-NMR spectroscopy makes the technique a valuable screening tool in the detection of inborn errors of purine and pyrimidine metabolism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Sigma-Aldrich
2′-Desoxyinosin, ≥98%