Merck
Adrenal steroid dysregulation in dystrophia myotonica.
Journal of internal medicine (1999-06-05)
A Johansson, A Henriksson, B O Olofsson, T Olsson
ZUSAMMENFASSUNG
To evaluate circulating adrenal steroid hormones, cortisol diurnal rhythm and the negative feedback function of the cortisol axis in patients with dystrophia myotonica (DyM), a disease where metabolic disturbances, peripheral insulin insensitivity and cognitive dysfunction are common features. Morning serum levels of dehydroepiandrosterone sulphate, androstenedione, 17 alpha-hydroxy progesterone and cortisol; morning serum levels of testosterone and insulin; diurnal rhythm of saliva cortisol; and an overnight dexamethasone suppression test, together with a cognitive screening test in men with DyM and in controls. Outpatient clinic in co-operation with Umeå University Hospital. Fifteen men with DyM and 13 age-matched controls. Adrenal steroid hormone levels, diurnal rhythm of saliva cortisol, dexamethasone suppression test and Mini Mental State Examination scores. Morning serum levels of dehydroepiandrosterone sulphate, androstenedione and 17 alpha-hydroxy progesterone were significantly decreased in DyM after inclusion of age and body mass index in multiple regression analyses (48, 26 and 32% decreases, respectively). An abnormal diurnal rhythm of saliva cortisol was present in all patients, mean saliva cortisol levels being significantly increased (33%) in DyM patients. Dexamethasone suppressibility did not differ between groups. DyM patients scored significantly lower on the Mini Mental State Examination (P < 0.001). These results indicate an abnormal adrenal steroid hormone secretion in DyM, which may contribute to peripheral insulin sensitivity as well as cognitive impairment in these patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Androstenedione solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
4-Androsten-3,17-dion, VETRANAL®, analytical standard