Merck
Maltose and isomaltose in uremic plasma following icodextrin administration.
Advances in peritoneal dialysis. Conference on Peritoneal Dialysis (2000-01-29)
R A Burke, J B Moberly, H Patel, T R Shockley, L Martis
ZUSAMMENFASSUNG
The presence of mixed disaccharides (maltose and isomaltose) in plasma from uremic patients has been previously investigated using gel-permeation chromatography. However, this method is unable to separate maltose (linked alpha-1-4) from isomaltose (linked alpha-1-6). We describe an alternative method using high-performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD) for the direct determination of maltose and isomaltose in uremic plasma. We measured maltose and isomaltose using HPAE-PAD in 6 normal subjects and in 15 uremic patients before and after once-daily icodextrin administration for at least 4 weeks. Both maltose and isomaltose were below limits of detection (< 1.0 mg/L) in plasma from normal controls. Patients with end-stage renal disease treated by continuous ambulatory peritoneal dialysis had elevated levels of isomaltose (23.6 +/- 8.3 mg/L) but low levels of maltose (< 3.0 mg/L). Treatment with icodextrin resulted in elevated plasma levels of maltose (range: 500-1600 mg/L), while levels of isomaltose declined to 9.8 +/- 5.2 mg/L (P < 0.0001 vs. baseline levels). We conclude that isomaltose (not maltose) is the primary disaccharide isomer that is elevated in the plasma of uremic patients, whereas maltose is the primary disaccharide isomer that is elevated following icodextrin administration. Furthermore, icodextrin administration results in an apparent reduction of isomaltose. Additional investigation will be required to address the mechanism for the reduction of isomaltose in patients treated by icodextrin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trimethylamin-N-oxid, 95%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Cadaverin, 95%
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Trimethylamin -Lösung, 43.0-49.0% in H2O (T)
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zink, foil, thickness 0.25 mm, 99.9% trace metals basis
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®