Merck
Dietary exposure estimates of 30 elements from the UK Total Diet Study.
Food additives and contaminants (2001-02-07)
G Ysart, P Miller, H Crews, P Robb, M Baxter, C De L'Argy, S Lofthouse, C Sargent, N Harrison
ZUSAMMENFASSUNG
Dietary exposures of consumers to 30 elements (aluminium, antimony, arsenic, barium, bismuth, boron, cadmium, calcium, chromium, cobalt, copper, germanium, gold, iridium, iron, lead, lithium, manganese, mercury, molybdenum, nickel, palladium, platinum, rhodium, ruthenium, selenium, strontium, thallium, tin and zinc) estimated from the UK 1994 Total Diet Study are reported, and compared with those from previous UK Total Diet Studies and those from other countries. Dietary exposure estimates were generally low and, where comparisons are possible, similar to those from other countries and below the relevant Provisional Tolerable Weekly Intakes and Provisional Maximum Tolerable Daily Intakes. For most of those elements included in previous UK Total Diet Studies, dietary exposures have declined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bismut, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismut, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismut, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismut, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismut, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismut, beads, 1-5 mm, 99.999% trace metals basis