Merck
Determination of trimebutine and desmethyl-trimebutine in human plasma by HPLC.
Arzneimittel-Forschung (2000-08-31)
M Lavit, S Saivin, H Boudra, F Michel, A Martin, G Cahiez, J P Labaune, J M Chomard, G Houin
ZUSAMMENFASSUNG
A simple and sensitive HPLC method has been developed to measure trimebutine (CAS 39133-31-8, maleate: CAS 34140-59-5) and its main metabolite desmethyl-trimebutine in human plasma. The method was validated according to the Washington Consensus Conference on the Validation of Analytical Methods. It involved extraction of the plasma with n-hexane containing 2-pentanol, followed by reversed-phase HPLC using a Partisil ODS2 10 microns column and UV detection at 265 nm. The retention times of the internal standard (procaine), desmethyl-trimebutine and trimebutine were 2.4, 4.3 and 6.5 min, respectively. The standard curves were linear from 20 ng.ml-1 (limit of quantitation) to 5000 ng.ml-1 for both compounds. The coefficient of variation for all the criteria of validation were less than 15%. The extraction recoveries obtained for trimebutine and desmethyl-trimebutine were about 90%. Both compounds were very stable upon storage in plasma. The method was tested by measuring the plasma concentrations following oral administration to humans during a bioequivalence study and was shown suitable for pharmacokinetic studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maleinsäure, ReagentPlus®, ≥99% (HPLC)
Supelco
Maleinsäure, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland