Merck
Behenic acid is a cholesterol-raising saturated fatty acid in humans.
The American journal of clinical nutrition (2000-12-22)
N B Cater, M A Denke
ZUSAMMENFASSUNG
Dietary behenic acid (22:0) is poorly absorbed. Because of its low bioavailability compared with other fatty acids and because of its very long chain length, the effect of dietary behenic acid (behenate) on serum lipid concentrations in humans is assumed to be neutral. The objective was to establish the cholesterol-raising potential of behenic acid by comparing the effects on lipid and lipoprotein concentrations of a specially formulated fat enriched with behenic acid with those of palm oil (rich in palmitic acid; 16:0) and high-oleic acid sunflower oil (rich in cis oleic acid; 18:1). In a randomized, crossover, metabolic-ward study, 7 mildly hypercholesterolemic men were fed 3 natural-food diets supplemented with behenate oil, palm oil, or high-oleic acid sunflower oil. Mean serum lipid and lipoprotein concentrations and plasma triacylglycerol fatty acid composition were determined from fasting blood drawn during the final 4 d of each 3-wk diet period. Behenate oil produced mean concentrations of total cholesterol (5.87+/-0.8 mmol/L) and LDL cholesterol (4.40+/-0.8 mmol/L) not significantly different from those produced by palm oil (5.84+/-0.7 and 4.42+/-0.7 mmol/L, respectively) but significantly higher than those produced by high-oleic acid sunflower oil (5.12+/-0.5 and 3.70+/-0.6 mmol/L, respectively). There were no significant differences in triacylglycerol or HDL-cholesterol concentrations. Despite its low bioavailability compared with oleic acid, behenic acid is a cholesterol-raising fatty acid in humans and is therefore not a suitable substitute for palmitic acid in manufactured triacylglycerols.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
Myristinsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Palmitinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Behensäure, 99%
Supelco
Oleinsäure, analytical standard
Supelco
Palmitinsäure, analytical standard
Sigma-Aldrich
Myristinsäure, natural, ≥98.5%, FG
Sigma-Aldrich
Palmitinsäure, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Myristinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Myristinsäure, ≥98.0% (GC)
Palmitinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Myristinsäure, analytical standard
Sigma-Aldrich
Palmitinsäure, natural, 98%, FG
Supelco
Behensäure, analytical standard
Supelco
Myristinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Oleinsäure, Selectophore, ≥99%