Merck
Effects of an oxalate load on urinary oxalate excretion in calcium stone formers.
Journal of renal nutrition : the official journal of the Council on Renal Nutrition of the National Kidney Foundation (2003-02-04)
Claudia de O G Mendonça, Ligia Araújo Martini, Alessandra Calábria Baxmann, José Luiz Nishiura, Lilian Cuppari, Dirce Maria Sigulem, Ita Pfeferman Heilberg
ZUSAMMENFASSUNG
To investigate the oxalate intake and the effect of an oxalate load on urinary oxalate excretion in calcium stone-forming (CSF) patients. Prospective study. University-affiliated outpatient Renal Lithiasis Unit. Seventy (70) CSF and 41 healthy subjects (HS) collected a 24-hour urine sample and were submitted to a 3-day dietary record to determine mean oxalate (Ox), calcium (Ca) and vitamin C intake. Fifty-eight (58) CSF patients were randomly selected to receive milk (N = 28) or dark (N = 30) chocolate as an oxalate load. Administration of either milk (94 mg Ox + 430 mg Ca) or dark chocolate (94 mg Ox + 26 mg Ca) for 3 days. A 24-hour urine sample was obtained before and after the load to determine calcium, oxalate, sodium, potassium, urea, and creatinine. Oxalate intake and excretion. CSF patients presented mean Ox intake of 98 +/- 137 mg/d, similar to that of HS (108 +/- 139 mg/d). Mean Ox and vitamin C intake was directly correlated with Ox excretion only in CSF. The consumption of dark chocolate induced a significant increase in mean urinary Ox (36 +/- 14 versus 30 +/- 10 mg/24 hr) not observed in the milk chocolate group. Thus, a 2-fold increase in Ox intake in this population of CSF patients produced a significant 20% increase in oxaluria, not observed when Ca was consumed simultaneously. The present study suggests that even small increases in Ox intake affect oxalate excretion and the mitigation of urinary oxalate increase by Ca consumption reinforces that Ca and Ox intakes for CSF patients should be in balance. Further studies are necessary to assess whether or not a 20% increase in oxaluria will lead to a higher risk of stone formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalsäure, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalsäure, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
Oxalsäure-Konzentrat, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC