Merck
  • Sensitive method for the quantification of urinary pyrimidine metabolites in healthy adults by gas chromatography-tandem mass spectrometry.
Sensitive method for the quantification of urinary pyrimidine metabolites in healthy adults by gas chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-06-12)
Ute Hofmann, Matthias Schwab, Sonja Seefried, Claudia Marx, Ulrich M Zanger, Michel Eichelbaum, Thomas E Mürdter
ZUSAMMENFASSUNG
Enzyme deficiencies in pyrimidine metabolism are associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil. To assess whether urinary levels of pyrimidines and their metabolites can be used for predicting patients' individual phenotype, a new gas chromatographic-tandem mass spectrometric method was developed which allows the simultaneous determination of uracil and thymine and their metabolites dihydrouracil, dihydrothymine, beta-ureidopropionic acid, beta-ureidoisobutyric acid, and the amino acids beta-alanine and beta-aminoisobutyric acid in human urine. Small aliquots (2-20 microl) of the urine samples were evaporated and derivatized to the tert.-butyldimethylsilyl derivatives before quantification, using the respective stable isotope-labelled analogues as internal standards. Analytical variation was acceptable with an intra-day imprecision (RSD) below 10%, for beta-ureidoisobutyric acid below 15%. The method was used for investigating the stability of urine samples and the influence of urine collection at different times.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dihydrouracil, powder
Sigma-Aldrich
3-Ureidopropionsäure, ≥98.0% (T)