Merck
Cobalt and antimony: genotoxicity and carcinogenicity.
Mutation research (2003-12-04)
Marlies De Boeck, Micheline Kirsch-Volders, Dominique Lison
ZUSAMMENFASSUNG
The purpose of this review is to summarise the data concerning genotoxicity and carcinogenicity of Co and Sb. Both metals have multiple industrial and/or therapeutical applications, depending on the considered species. Cobalt is used for the production of alloys and hard metal (cemented carbide), diamond polishing, drying agents, pigments and catalysts. Occupational exposure to cobalt may result in adverse health effects in different organs or tissues. Antimony trioxide is primarily used as a flame retardant in rubber, plastics, pigments, adhesives, textiles, and paper. Antimony potassium tartrate has been used worldwide as an anti-shistosomal drug. Pentavalent antimony compounds have been used for the treatment of leishmaniasis. Co(II) ions are genotoxic in vitro and in vivo, and carcinogenic in rodents. Co metal is genotoxic in vitro. Hard metal dust, of which occupational exposure is linked to an increased lung cancer risk, is proven to be genotoxic in vitro and in vivo. Possibly, production of active oxygen species and/or DNA repair inhibition are mechanisms involved. Given the recently provided proof for in vitro and in vivo genotoxic potential of hard metal dust, the mechanistic evidence of elevated production of active oxygen species and the epidemiological data on increased cancer risk, it may be advisable to consider the possibility of a new evaluation by IARC. Both trivalent and pentavalent antimony compounds are generally negative in non-mammalian genotoxicity tests, while mammalian test systems usually give positive results for Sb(III) and negative results for Sb(V) compounds. Assessment of the in vivo potential of Sb2O3 to induce chromosome aberrations (CA) gave conflicting results. Animal carcinogenicity data were concluded sufficient for Sb2O3 by IARC. Human carcinogenicity data is difficult to evaluate given the frequent co-exposure to arsenic. Possible mechanisms of action, including potential to produce active oxygen species and to interfere with DNA repair systems, still need further investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Antimon, powder, -100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Kobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Antimon, beads, ≤5 mm, low oxide, 99.999% trace metals basis
Sigma-Aldrich
Kobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Kobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Kobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Kobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Antimon, rod, 6mm, diameter 6mm, 99.999%
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, ≥99.99%
Antimon, rod, 20mm, diameter 4.7mm, 99.999%
Sigma-Aldrich
Kobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Antimon, rod, 15mm, diameter 2.0mm, 99.99%
Antimon, rod, 50mm, diameter 2.0mm, 99.99%
Antimon, rod, 25mm, diameter 2.0mm, 99.99%
Antimon, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
Antimon, rod, 50mm, diameter 4.7mm, 99.999%
Antimon, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 95+%
Antimon, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 95+%
Antimon, microfoil, 50x50mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
Antimon, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 95+%
Antimon, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 95+%
Antimon, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 95+%
Antimon, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 95+%
Antimon, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 95+%
Antimon, microfoil, 25x25mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
Antimon, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 95+%
Antimon, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 95+%