Merck
Plasma and dietary carotenoids, and the risk of prostate cancer: a nested case-control study.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2004-02-20)
Kana Wu, John W Erdman, Steven J Schwartz, Elizabeth A Platz, Michael Leitzmann, Steven K Clinton, Valerie DeGroff, Walter C Willett, Edward Giovannucci
ZUSAMMENFASSUNG
The association between plasma carotenoids and prostate cancer risk was investigated in a case-control study nested within the prospective Health Professionals Follow-up Study. We matched 450 incident prostate cancer cases diagnosed from 1993-1998 to 450 controls by age, time, month, and year of blood donation. Modest inverse, but not statistically significant, associations were observed among plasma alpha-carotene, beta-carotene, and lycopene concentrations, and overall risk of prostate cancer diagnosis [odds ratio (highest versus lowest quintile; OR), alpha-carotene: OR, 0.67 [95% confidence interval (CI), -0.40-1.09]; beta-carotene: OR, 0.78 (95% CI, 0.48-1.25); lycopene: OR, 0.66 (95% CI, 0.38-1.13)]. The inverse association between plasma lycopene concentrations and prostate cancer risk was limited to participants who were 65 years or older (OR, 0.47; 95% CI, 0.23-0.98) and without a family history of prostate cancer (OR, 0.48; 95% CI, 0.26-0.89). Combining, older age and a negative family history provided similar results (OR, 0.43; 95% CI, 0.18-1.02). Inverse associations between beta-carotene and prostate cancer risk were also found among younger participants (<65 years of age; OR, 0.36; 95% CI, 0.14-0.91; P(trend) = 0.03). Combining dietary intake and plasma data confirmed our results. We found a statistically significant inverse association between higher plasma lycopene concentrations and lower risk of prostate cancer, which was restricted to older participants and those without a family history of prostate cancer. This observation suggests that tomato products may exhibit more potent protection against sporadic prostate cancer rather than those with a stronger familial or hereditary component. In addition, our findings also suggest that among younger men, diets rich in beta-carotene may also play a protective role in prostate carcinogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Carotin, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
β-Carotin, synthetic, ≥93% (UV), powder
Supelco
β-Carotin, Pharmaceutical Secondary Standard; Certified Reference Material