Merck
  • Hepatocyte proliferation in chronic hepatitis C: correlation with degree of liver disease and serum alpha-fetoprotein.
Hepatocyte proliferation in chronic hepatitis C: correlation with degree of liver disease and serum alpha-fetoprotein.
Liver international : official journal of the International Association for the Study of the Liver (2004-06-11)
P Wilfredo Canchis, Stevan A Gonzalez, M Isabel Fiel, Luis Chiriboga, Herman Yee, Brian R Edlin, Ira M Jacobson, Andrew H Talal
ZUSAMMENFASSUNG
Hepatocyte proliferation (HP) is an adaptive response to liver injury. The relationships between HP and necroinflammation, fibrosis, and serum alpha-fetoprotein (AFP) levels in chronic hepatitis C virus (HCV) infection, however, are not well understood. Proliferative hepatocytes (Ki-67+) were identified using immunohistochemical staining in formalin-fixed, paraffin-embedded liver tissue from 156 HCV RNA-positive patients with different degrees of liver histopathology. Twenty high-power fields (HPFs) in lobular areas were counted in each specimen. HP increased by 1.22 +/- 0.25 cells/HPF per increase in necroinflammation from grade 0 (median: 0.13; range: [0.1-0.5] cells/HPF) through grade 3 (median: 1.80; range: [0.0-25.2] cells/HPF; P=0.002). HP increased by 0.81 +/- 0.20 cells/HPF per increase in fibrosis from stage 0 (median: 0.33; range: [0.0-1.3] cells/HPF) through stage 3 (median: 1.70; range: [0.0-25.2] cells/HPF) and then decreased in stage 4 (to median: 0.90; range: [0.0-5.3] cells/HPF). HP also increased with advancing age (P=0.03). Among patients with advanced liver disease, HP was no higher in patients with elevated serum AFP levels (median: 1.68; range: [0.1-5.3] cells/HPF) than in those with normal serum AFP levels (median: 1.70; range: [0.0-25.2] cells/HPF; P=0.26). In patients with chronic HCV infection, HP increases with histologic progression of liver disease, but is impaired in cirrhosis. HP was not increased in patients with elevated serum AFP levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.