Merck
Significance of urinary tartaric acid.
Clinical chemistry (2005-03-02)
Richard S Lord, Cheryl K Burdette, J Alexander Bralley
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(+)-Tartarsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
L-(+)-Tartarsäure, ≥99.5%
Sigma-Aldrich
L-(+)-Tartarsäure, ≥99.7%, FCC, FG
Sigma-Aldrich
L-(+)-Tartarsäure, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
L-(+)-Tartarsäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
L-(+)-Tartarsäure, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
L-(+)-Tartarsäure, BioXtra
Supelco
L-(+)-Tartarsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland