Merck
  • Effects of nonselective cyclooxygenase inhibition with low-dose ibuprofen on tumor growth, angiogenesis, metastasis, and survival in a mouse model of colorectal cancer.
Effects of nonselective cyclooxygenase inhibition with low-dose ibuprofen on tumor growth, angiogenesis, metastasis, and survival in a mouse model of colorectal cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2005-03-05)
Min Yao, Wei Zhou, Simren Sangha, Andrew Albert, Albert J Chang, Thomas C Liu, M Michael Wolfe
ZUSAMMENFASSUNG
To determine whether the nonselective and relatively inexpensive nonsteroidal anti-inflammatory drug ibuprofen would be effective in inhibiting colorectal cancer and might improve mortality in a mouse model. The effects of ibuprofen on tumor growth inhibition and animal survival have been examined in both mouse and human colorectal cancer tumor models. Angiogenesis was measured by in vitro endothelial cell tube formation and immunohistochemistry. Ibuprofen significantly inhibited cell proliferation in mouse (MC-26) and human (HT-29) colorectal cancer cell lines. In vitro angiogenesis assays also indicated that ibuprofen decreased both cell proliferation and tube formation. The administration of chow containing 1,360 ppm ibuprofen, which achieved an average plasma concentration of ibuprofen lower than the peak level achieved in humans at therapeutic doses, inhibited tumor growth by 40% to 82%. Fewer liver metastases were found in the ibuprofen group compared with the control group. In combination therapy with the standard antineoplastic agents, 5-fluorouracil, or irinotecan (CPT-11), tumor volumes in the groups with ibuprofen +/- CPT-11 or 5-fluorouracil were smaller than in the control group. Ibuprofen was similar to the cyclooxygenase-2 selective inhibitor rofecoxib in its ability to suppress tumor growth and improve overall survival. Ibuprofen, in part by modulating tumor angiogenesis, decreases both tumor growth and metastatic potential in mice. The ibuprofen doses were in the low range of therapeutic human plasma concentrations. Ibuprofen potentiates the antitumor properties of CPT-11 and improves survival of mice without increasing gastrointestinal toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ibuprofen, meets USP testing specifications