Merck
  • Delayed effects of short-term transdermal application of 7-oxo-dehydroepiandrosterone on its metabolites, some hormonal steroids and relevant proteohormones in healthy male volunteers.
Delayed effects of short-term transdermal application of 7-oxo-dehydroepiandrosterone on its metabolites, some hormonal steroids and relevant proteohormones in healthy male volunteers.
Clinical chemistry and laboratory medicine (2005-04-22)
Jarmila Sulcová, Richard Hampl, Martin Hill, Luboslav Stárka, Alois Novácek
ZUSAMMENFASSUNG
Twenty-one healthy male volunteers aged 20-70 years were given transdermally 25 mg of 7-oxo-dehydroepiandrosterone daily in the form of an emulgel for 8 consecutive days. Morning blood was collected as follows: before application, and after the first, fourth and eighth doses (days 0, 2, 5 and 9), and then at different time intervals after termination of the treatment (days 16, 23, 37, 51, 72 and 100). Cortisol, testosterone, epitestosterone, estradiol, dehydroepiandrosterone and its sulfate, 7alpha- and 7beta-hydroxy-dehydroepiandrosterone, luteinizing hormone, follicle-stimulating hormone and sex hormone-binding globulin were measured in blood sera. In the course of treatment 7beta-hydroxy-dehydroepiandrosterone was significantly increased; testosterone and gonadotropins were lowered, but only after the first dose. All other significant changes were observed during the period after termination of the application:7beta-hydroxy-dehydroepiandrosterone remained increased for 28 days, 7alpha-hydroxy-dehydroepiandrosterone, testosterone, estradiol and sex hormone-binding globulin were decreased as late as day 63 and 91, respectively. On the other hand, epitestosterone was significantly increased between days 23 and 100. The levels of all other parameters studied were not significantly changed. The study points to an immediate as well as delayed effect of the short-term transdermal application of 7-oxo-dehydroepiandrosterone on relevant hormonal parameters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
17β-Estradiol -Lösung, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, analytical standard