Merck
  • Effects of aluminum on the nervous system and its possible link with neurodegenerative diseases.
Effects of aluminum on the nervous system and its possible link with neurodegenerative diseases.
Journal of Alzheimer's disease : JAD (2005-11-26)
Masahiro Kawahara
ZUSAMMENFASSUNG
Aluminum is environmentally abundant, but not an essential element. Aluminum has been associated with several neurodegenerative diseases, such as dialysis encephalopathy, amyotrophic lateral sclerosis and Parkinsonism dementia in the Kii peninsula and Guam, and in particular, Alzheimer's disease. Although this association remains controversial, there is increasing evidence which suggests the implication of metal homeostasis in the pathogenesis of Alzheimer's disease. Aluminum, zinc, copper, and iron cause the conformational changes of Alzheimer's amyloid-beta protein. Al causes the accumulation of tau protein and amyloid-beta protein in experimental animals. Aluminum induces neuronal apoptosis in vivo as well as in vitro. Furthermore, a relationship between aluminum and the iron-homeostasis or calcium-homeostasis has been suggested. Based on these findings, the characteristics of aluminum neurotoxicity are reviewed, and the potential link between aluminum and neurodegenerative diseases is reconsidered.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminium, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminium, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminium, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminium, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminium, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminium, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminium, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminium, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminium, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminium, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminium, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminium, foil, thickness 1.0 mm, 99.999% trace metals basis
Aluminium, IRMM®, certified reference material, 1.0 mm foil
Aluminium, IRMM®, certified reference material, 0.1 mm foil
Aluminium, IRMM®, certified reference material, 1.0 mm wire