Merck
  • Ezetimibe for the treatment of uncontrolled hypercholesterolemia in patients with high-dose statin therapy after renal transplantation.
Ezetimibe for the treatment of uncontrolled hypercholesterolemia in patients with high-dose statin therapy after renal transplantation.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2006-01-26)
M Kohnle, F Pietruck, A Kribben, Th Philipp, U Heemann, O Witzke
ZUSAMMENFASSUNG
We investigated prospectively the efficacy of ezetimibe in addition to statin therapy in stable renal transplant patients in whom hypercholesterolemia was not sufficiently treated. Eighteen renal transplant patients received 10 mg ezetimibe once daily in addition to high-dose statin therapy for uncontrolled hypercholesterolemia. Total cholesterol, LDL-cholesterol, HDL-cholesterol, triglycerides, Tacrolimus (Tac)- and Cyclosporine A (CsA) blood levels, creatinine, urea, liver enzymes, electrolytes and creatinkinase (CK) were measured before initiation of ezetimibe therapy, after 7 days, 6 weeks and 3 months. Cholesterol concentrations decreased significantly (p < 0.005) from 264 +/- 46 mg/dL at baseline to 205 +/- 48 mg/dL after 1 week to 202 +/- 48 mg/dL after 6 weeks and 212 +/- 40 mg/dL after 3 months (reduction after 3 months 21 +/- 10%). LDL-concentrations decreased significantly (p < 0.005) from 178 +/- 41 mg/dL at baseline to 129 +/- 35 mg/dL after 1 week to 123 +/- 25 after 6 weeks and to 117 +/- 40 mg/dL after 3 months (reduction after 3 months 37 +/- 14%). Two patients stopped ezetimibe therapy due to nausea and muscle pain without CK elevation. Significant changes of CsA and Tac blood levels, liver and muscle enzymes were not observed. Ezetimibe seems to be an effective therapy for uncontrolled hypercholesterolemia in renal transplant patients when combined with high-dose statin therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Harnstoff, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C