Merck
  • Formation of epoxyeicosatrienoic acids from arachidonic acid by cultured rat aortic smooth muscle cell microsomes.
Formation of epoxyeicosatrienoic acids from arachidonic acid by cultured rat aortic smooth muscle cell microsomes.
Prostaglandins, leukotrienes, and essential fatty acids (1991-03-01)
K Hasunuma, T Terano, Y Tamura, S Yoshida
ZUSAMMENFASSUNG
The vasodilatory effect of epoxyeicosatrienoic acids (EpETrE), especially 5(6)-EpETrE, has been reported recently and a role of P-450-dependent arachidonic acid monooxygenase metabolites was suggested in vasoregulation. Accordingly, the presence of P-450-dependent arachidonic acid monooxygenase was investigated in rat aortic smooth muscle cells. Incubation of the microsomes of rat cultured aortic smooth muscle cells with 14C-arachidonic acid in the presence of 1 mM NADPH resulted in the formation of oxygenated metabolites. The metabolites were separated and purified by reverse phase and straight phase high performance liquid chromatography and identified by gas chromatography-mass spectrometry. Identified metabolites were 5(6)-EpETrE, 5,6-dihydroxyeicosatrienoic acid (DiHETrE), and 14,15-DiHETrE. The formation of these metabolites was totally dependent on the presence of NADPH, and inhibitors of cytochrome P-450-dependent enzymes, SKF-525A and metyrapone, reduced the formation of these metabolites. This is the first report that cytochrome P-450-dependent arachidonic acid metabolites, especially 5(6)-EpETrE and 14(15)-EpETrE, can be produced in the microsomes of vascular smooth muscle cells of rats.