Merck
  • Coenzyme Q10 increases T4/T8 ratios of lymphocytes in ordinary subjects and relevance to patients having the AIDS related complex.
Coenzyme Q10 increases T4/T8 ratios of lymphocytes in ordinary subjects and relevance to patients having the AIDS related complex.
Biochemical and biophysical research communications (1991-05-10)
K Folkers, T Hanioka, L J Xia, J T McRee, P Langsjoen
ZUSAMMENFASSUNG
Coenzyme Q10 (CoQ10) is indispensable to biochemical mechanisms of bioenergetics, and it has a non-specific role as an antioxidant. CoQ10 has shown a hematological activity for the human and has shown an influence on the host defense system. The T4/T8 ratios of lymphocytes are known to be low in patients with AIDS, ARC and malignancies. Our two patients with ARC have survived four-five years without any symptoms of adenopathy or infection on continuous treatment with CoQ10. We have newly found that 14 ordinary subjects responded to CoQ10 by increases in the T4/T8 ratios and an increase in blood levels of CoQ10; both by p less than 0.001. This knowledge and survival of two ARC patients for four-five years on CoQ10 without symptoms, and new data on increasing ratios of T4/T8 lymphocytes in the human by treatment with CoQ10 constitute a rationale for new double blind clinical trials on treating patients with AIDS, ARC and diverse malignancies with CoQ10.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Coenzym Q10, ≥98% (HPLC)