Merck
Evaluation of the preruminant calf as a model for the study of human carotenoid metabolism.
The Journal of nutrition (1992-02-11)
C L Poor, T L Bierer, N R Merchen, G C Fahey, M R Murphy, J W Erdman
ZUSAMMENFASSUNG
This study evaluated the preruminant calf as an animal model for the study of human carotenoid metabolism. Fifteen newborn male Holstein calves were fed a carotenoid-free milk replacer diet to maintain them in the preruminant state. After a 7-d adjustment period, three calves were killed and 12 calves received a single oral dose (20 mg) of beta-carotene in the form of water-soluble beadlets. Blood samples were collected periodically and samples of various tissues were collected when the calves were killed. Three animals each were killed by exsanguination at 1, 3, 6 and 11 d post-dosing. Serum beta-carotene concentrations peaked between 12 and 30 h post-dosing and declined slowly afterwards. Serum data were fitted to a two-compartment model and yielded an elimination constant (k2) that was similar to reported human values. Adrenal tissue showed significant concentrations of beta-carotene by 24 h post-dosing, and levels were still elevated at 264 h. Liver, spleen and lung beta-carotene concentrations were significantly elevated by 24 h and rapidly declined thereafter. Adipose and kidney peak beta-carotene concentrations were observed at 72 and 144 h, respectively. Heart and muscle did not display significant changes in beta-carotene concentrations. The preruminant calf shows promise as an animal model for the study of absorption and metabolism of carotenoids by humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Carotin, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
β-Carotin, synthetic, ≥93% (UV), powder
Supelco
β-Carotin, Pharmaceutical Secondary Standard; Certified Reference Material