Merck
Glycolate determination detects type I primary hyperoxaluria in dialysis patients.
Kidney international (1991-01-01)
M Marangella, M Petrarulo, O Bianco, C Vitale, P Finocchiaro, F Linari
ZUSAMMENFASSUNG
The detection of type I primary hyperoxaluria is based on the finding of exceedingly high oxalate excretion which is associated with increased glycolate excretion. The differential diagnosis of this disease may become a difficult task once end-stage renal disease (ESRD) and anuria have supervened. The various procedures thus far proposed to obviate this circumstance are complex, inaccurate or not reproducible. In this paper we propose the accurate liquid chromatographic determination of glycolate in blood and dialysate as a means to detect type I primary hyperoxaluria in patients on maintenance hemodialysis (RDT). The method is based on the enzymatic conversion of glycolate to glyoxylate coupled with alpha-keto acid derivatization with phenylhydrazine. The resulting phenylhydrazone is then resolved by high-performance liquid chromatograph (HPLC). With this method, plasma glycolate in 12 healthy controls was 7.8 +/- 1.7 mumol/liter, almost twentyfold less than previously reported. Five dialysis patients with high serum oxalate, of whom four with primary hyperoxaluria and one with Crohn's disease and presumed enteric oxalate hyperabsorption, were checked by this method and compared to nine patients with oxalosis-unrelated ESRD. The patients with hyperoxalemia were also evaluated for their response to pyridoxine therapy. The measurement of glycolate in blood drawn prior to and at the end of the dialysis session as well as in the dialysate soundly discriminated the patients with type I hyperoxaluria from all the other dialysis patients. Glycolate measurement was shown to be much more powerful than oxalate in that patients with oxalosis-induced ESRD exhibited an almost two hundred and fiftyfold increase compared to the oxalosis-unrelated patients.(ABSTRACT TRUNCATED AT 250 WORDS)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycolsäure, ReagentPlus®, 99%
Sigma-Aldrich
Glycolsäure -Lösung, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolsäure -Lösung, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolsäure, BioXtra, ≥98.0% (titration)
Supelco
Glycolsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland