Merck
  • Monoamine metabolites and amino acids in serum from schizophrenic patients before and during sulpiride treatment.
Monoamine metabolites and amino acids in serum from schizophrenic patients before and during sulpiride treatment.
Psychopharmacology (1989-01-01)
G Alfredsson, F A Wiesel
ZUSAMMENFASSUNG
Twenty-four acutely ill schizophrenic patients (DSM-III-R), 18-42 years old, were treated for 6 weeks with sulpiride. Sulpiride was administered in three different daily dosages (starting with 400, 800 or 1200 mg) according to a double blind randomized administration schedule. The monoamine metabolites (MAM) homovanillic acid (HVA), 5-hydroxy-indoleacetic acid (5-HIAA), 4-hydroxy-3-methoxy-phenylglycol (HMPG) and the amino acids tyrosine, tryptophan, glutamate and glutamine were measured in serum before treatment and once a week during treatment. There were no significant differences between healthy controls and schizophrenic patients in serum levels of monoamine metabolites and amino acids before treatment. There was no dose-response effect of sulpiride on serum levels of the monoamine metabolites or the amino acids. The results are therefore based on the whole group of patients. During treatment the HMPG levels were reduced at all points in time. The serum level of HVA was significantly reduced after 6 weeks. The 5-HIAA and the amino acid levels were not changed during treatment. There were no significant correlations among the monoamine metabolites before treatment. During treatment, however, significant correlations were found among MAM and amino acids. Since the biochemical findings during the treatment were not related to the dose or the concentration of sulpiride the results may be related to secondary biochemical effects of sulpiride and/or to changes in the clinical state following treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutaminlösung 200 mmol/l, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Asparaginsäure, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Glutaminsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Taurin, ≥99%
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Isoleucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Glutamin
Sigma-Aldrich
L-Glutaminsäure, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Phenylalanin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
Sigma-Aldrich
L-Asparagin, ≥98% (HPLC)
Sigma-Aldrich
Taurin, suitable for cell culture, meets USP testing specifications
SAFC
L-Methionin
Sigma-Aldrich
L-Citrullin, ≥98% (TLC)
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Methionin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Asparagin, BioReagent, suitable for cell culture, suitable for insect cell culture