Merck
  • Acyl derivatives of p-aminobenzoic acid as new substrates for the assay of serum acylase activity.
Acyl derivatives of p-aminobenzoic acid as new substrates for the assay of serum acylase activity.
Clinica chimica acta; international journal of clinical chemistry (1978-03-01)
A Szewczuk, M Wellman-Bednawska
PMID305833
ZUSAMMENFASSUNG
A simple colorimetric method for the assay of cobalt-activated acylase activity in human serum using new and less toxic N-chloroacetyl-gamma-L-glutamyl-p-aminobenzoic acid as substrate has been described. The values obtained with this method are almost the same as with the previously described method using naphthylamide substrate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Aminobenzoesäure, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Aminobenzoesäure, ReagentPlus®, 99%
Supelco
4-Aminobenzoesäure, analytical standard
Sigma-Aldrich
4-Aminobenzoesäure, purified by sublimation, ≥99%