- [Use of nicotinic acid and midocalm for correcting blood coagulation in patients with radiation edema of the extremities].
[Use of nicotinic acid and midocalm for correcting blood coagulation in patients with radiation edema of the extremities].
Meditsinskaia radiologiia (1985-08-01)
S I Chekalina, L I Guseva, M S Bardyechev
PMID4033386
ZUSAMMENFASSUNG
A study was made of the functional activity of platelets, plasmatic coagulation and fibrinolysis in patients with late radiation disorders of the blood and lymph flow during treatment with vasodilative drugs: nicotinic acid and midocalm. These drugs tend to decrease the functional activity of platelets and to raise the blood fibrinolytic activity. Their use is indicated to this group of patients to better their intravascular status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%