Merck
Semiautomated enzymic microassay for plasma L-alanine.
Enzyme (1979-01-01)
A Stahl, A Frick, M Imler, J L Schlienger
PMID41707
ZUSAMMENFASSUNG
Plasma L-alanine is deaminated by bacterial alanine-dehydrogenase; the resulting ammonia is dialyzed out and measured by use of a continuous flow phenol-hypochlorite colorimetric microassay. Concentrations in the range of 10-1,000 mumol alanine/l can be determined in a 50-microliter sample. The optimal conditions for the assay are specified. Study of the analytical qualities of the technique shows high specificity, good reproducibility , and a detection limit of 6 mumol/l. Usual values in human plasma from arterial or venous blood are respectively 296 +/- 166 and 376 +/- 214 mumol alanine/l (x +/- 2 SD). The usual values in rats are close to those found in man.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Alanin, ≥98% (TLC)
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanin, ≥99%
Supelco
L-Alanin, Pharmaceutical Secondary Standard; Certified Reference Material
Alanin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanin-12C3, 99.9 atom % 12C