Merck
[Contents of putrescine, spermidine and spermine in tissue of the human brain glial tumors].
Ukrainskii biokhimicheskii zhurnal (1978) (1979-09-01)
P A Mirzoian, M Sh Promyslov
PMID516181
ZUSAMMENFASSUNG
It is shown that in the tissue of the human brain glial tumours the content of putrescine depends on the degree of the tumour malignization. In malignant gliomas (glioblastomas), as compared to the benign (astrocytomas), the content of putrescine is significantly higher. The content of spermidine in glial tumours of a malignancy different degree is twice as high as the level of this polyamine in the brain grey matter, and it is twice as low as in the white matter. The content of spermine in the brain glial tumours does not differ essentially from its level in the brain tissue.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Spermidin, ≥99% (GC)
Sigma-Aldrich
Spermidin, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Spermidin, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Spermidin 0,1 mol/l -Lösung
Supelco
Spermidin, analytical standard