Merck
The influence of long-term treatment with timolol on human tear lysozyme albumin content.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (1982-01-01)
G Strasser, G Grabner
ZUSAMMENFASSUNG
The long-term effect of timolol maleate on lacrimal secretion was tested by investigating the concentration of lysozyme and albumin in tears before the start of treatment and after different follow-up periods. A total of 25 patients (45 eyes) were treated with 0.25% or 0.5% timolol as the only antiglaucomatous therapy over a 7.6 month mean observation period (2-13 months). No significant change was found either in the lysozyme or in the albumin concentration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maleinsäure, ReagentPlus®, ≥99% (HPLC)
Supelco
Maleinsäure, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland