Merck
  • Tissue distribution of cocaine methyl esterase and ethyl transferase activities: correlation with carboxylesterase protein.
Tissue distribution of cocaine methyl esterase and ethyl transferase activities: correlation with carboxylesterase protein.
The Journal of pharmacology and experimental therapeutics (1995-11-01)
R A Dean, J Zhang, M R Brzezinski, W F Bosron
ZUSAMMENFASSUNG
The tissue distribution of cocaine methyl esterase and ethanol-dependent ethyl transferase activities was determined in the rat and compared to the tissue distribution of three distinct non-specific hydrolases. Rates of formation of benzoylecgonine from cocaine and cocaethylene from ethanol and cocaine were measured in serum and tissue homogenate-supernatants of the brain, heart, kidney, liver, lung and spleen. The tissue distribution of three nonspecific esterases, A, B and C, was defined by nondenaturing gel electrophoresis and measuring the hydrolysis of 4-methylumbelliferyl acetate in the gels. Immunoreactive protein was localized by using Western blot analysis with polyclonal rabbit antihuman liver cocaine methyl esterase antibody after denaturing and nondenaturing gel electrophoresis. The rat liver, lung, kidney and heart exhibited cocaine methyl esterase and ethyl transferase activities and immunoreactive protein. The brain had cocaine methyl esterase activity but no ethyl transferase activity; neither activity was found in serum or spleen. The dominant immunoreactive bands in the liver, lung, kidney and heart comigrated with the 59 kD band of purified human liver cocaine methyl esterase. The rat liver, lung and kidney exhibited a band of nonspecific esterase activity that migrated with purified human liver cocaine methyl esterase and rat hydrolase A. These observations suggest that rat hydrolase A is similar to human cocaine methyl esterase. The lack of straight forward correlation between cocaine methyl esterase activity and immunoreactive protein and nonspecific esterase activity suggests that more than one enzyme catalyzes the hydrolysis of cocaine to benzoylecgonine in the rat.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 Ampullen/Kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 Ampullen/Kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®