Merck
  • Reduced and oxidized forms of glutathione and alpha-tocopherol in the cerebrospinal fluid of parkinsonian patients: comparison between before and after L-dopa treatment.
Reduced and oxidized forms of glutathione and alpha-tocopherol in the cerebrospinal fluid of parkinsonian patients: comparison between before and after L-dopa treatment.
Neuroscience letters (1995-01-16)
H Tohgi, T Abe, M Saheki, F Hamato, K Sasaki, S Takahashi
ZUSAMMENFASSUNG
In the cerebrospinal fluid of untreated patients with Parkinson's disease (PD) the concentrations of reduced glutathione (GSH) and alpha-tocopherol (alpha-TOH) were unaltered but the concentration of oxidized glutathione (glutathione disulfide, GSSG) (P < 0.001), the GSSG/GSH ratio (P < 0.06), alpha-tocopherol quinone (alpha-TQ) (P < 0.001), and the alpha-TQ/alpha-TOH ratio (P < 0.01) were reduced significantly. In L-dopa-treated patients, the concentrations of GSH, GSSG, and the alpha-TQ concentration and the alpha-TQ/alpha-TOH ratio (P < 0.05) increased compared with untreated PD patients. These results suggest that oxidation of GSH and alpha-TOH is decreased in untreated PD patients, but is activated to a control level or more after L-dopa treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion, oxidiert, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathion, oxidiert, ≥98%, lyophilized powder
Sigma-Aldrich
L-Glutathion, oxidiert, BioXtra, ≥98%