Merck
Reference ranges for serum cortisol and 17-hydroxyprogesterone levels in preterm infants.
The Journal of pediatrics (1995-06-01)
S al Saedi, H Dean, W Dent, C Cronin
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
17α-Hydroxyprogesteron, ≥95%
Supelco
17α-Hydroxyprogesterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®