Merck
  • Inelastic electron tunneling spectroscopic study of biological compounds in human sweat adsorbed on alumina.
Inelastic electron tunneling spectroscopic study of biological compounds in human sweat adsorbed on alumina.
Analytical chemistry (1994-03-15)
M Higo, S Kamata
ZUSAMMENFASSUNG
Inelastic electron tunneling spectroscopy has been used for the detection and characterization of biological compounds in human sweat adsorbed on alumina surfaces. The sweat was collected from students and distributed to aqueous and benzene phases by solvent extraction. Both solutions were doped onto alumina surfaces of tunneling junctions, and the vibrational spectra of the adsorbed species were measured by inelastic electron tunneling spectroscopy. Analysis of these tunneling spectra showed that lactic acid in the sweat is extracted with water and selectively adsorbed on alumina. Adsorbed fatty acids were detected from the benzene phase. These biological compounds in the sweat solutions were found to react with the alumina to give about a monolayer of adsorbed carboxylate anions on the surfaces.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzylalkohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzylalkohol, ACS reagent, ≥99.0%
Supelco
Benzylalkohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzylalkohol, anhydrous, 99.8%
Sigma-Aldrich
Benzylalkohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzylalkohol, ≥99%, FCC, FG
Supelco
Benzylalkohol, analytical standard
Sigma-Aldrich
Benzylalkohol, natural, ≥98%, FG
Benzylalkohol, European Pharmacopoeia (EP) Reference Standard