- Automated screening system for purine and pyrimidine metabolism disorders using high-performance liquid chromatography.
Automated screening system for purine and pyrimidine metabolism disorders using high-performance liquid chromatography.
Journal of chromatography. B, Biomedical applications (1995-10-20)
S Sumi, K Kidouchi, S Ohba, Y Wada
PMID8581129
ZUSAMMENFASSUNG
An automated screening system for purine and pyrimidine metabolism disorders using high-performance liquid chromatography (HPLC) with column switching is described. The system consists of a reversed-phase column, a cation-exchange column, a column switch, four sets of ultraviolet absorbance detectors, a microcomputer and other conventional equipment. As this system permits the simultaneous determination of urinary orotic acid, uracil, dihydrouracil, pseudouridine, xanthine, 2,8-dihydroxyadenine and succinyladenosine, it offers a useful method for the detection of orotic aciduria, dihydropyrimidine dehydrogenase deficiency, dihydropyrimidinuria, xanthinuria, adenine phosphoribosyltransferase deficiency and adenylosuccinase deficiency.