Merck
  • MDC-9 (ADAM-9/Meltrin gamma) functions as an adhesion molecule by binding the alpha(v)beta(5) integrin.
MDC-9 (ADAM-9/Meltrin gamma) functions as an adhesion molecule by binding the alpha(v)beta(5) integrin.
Biochemical and biophysical research communications (2001-02-13)
M Zhou, R Graham, G Russell, P I Croucher
ZUSAMMENFASSUNG
MDC-9 is a widely expressed member of the metalloproteinase/disintegrin/cysteine-rich protein family. The disintegrin domain of MDC-9 lacks an RGD motif but has recently been reported to bind the alpha(6)beta(1) integrin; however, it is unclear whether MDC-9 can bind other integrins. In the present study myeloma cells, but not lymphoblastoid cells, were shown to bind to immobilised, recombinantly expressed MDC-9 disintegrin domain (A9dis). Binding was divalent cation-dependent, being supported by Mn(2+) and Ca(2+). Adhesion of myeloma cells to A9dis was completely inhibited by an antibody to the alpha(v)beta(5) integrin but not by antibodies to other subunits. RGD-containing peptides had no effect on binding, suggesting that MDC-9 interacts with alpha(v)beta(5) in an RGD-independent manner. Flow cytometric analyses demonstrated that myeloma cells, but not lymphoblastoid cells, expressed alpha(v)beta(5) on the cell membrane. These data indicated that the disintegrin domain of MDC-9 can function as an adhesion molecule by interacting with an alpha(v)beta(5) integrin.