Merck
  • Method for determination of stable carbon isotope ratio of methylnitrophenols in atmospheric particulate matter.
Method for determination of stable carbon isotope ratio of methylnitrophenols in atmospheric particulate matter.
Atmospheric Measurement Techniques (2011)
Moukhtar S, et al.