- Construction of a full-length enriched and a 5'-end enriched cDNA library using the oligo-capping method.
Construction of a full-length enriched and a 5'-end enriched cDNA library using the oligo-capping method.
Methods in molecular biology (Clifton, N.J.) (2003-04-22)
Yutaka Suzuki, Sumio Sugano
PMID12703735