Merck
  • Use of nitroanilines for spectrophotometric determination of ethinylestradiol in pharmaceutical formulations.
Use of nitroanilines for spectrophotometric determination of ethinylestradiol in pharmaceutical formulations.
Analytical Methods : Advancing Methods and Applications (2011)
Leonardo SG, et al.