Merck
  • Permselectivity, solubility and diffusivity of propyl propionate/water mixtures in poly (ether block amide) membranes.
Permselectivity, solubility and diffusivity of propyl propionate/water mixtures in poly (ether block amide) membranes.
Journal of Membrane Science (2007)
Mujiburohman M and Feng X.