Merck
  • Mechanistic Aspects of the Electrochemical Oxidation of Diethyl 1, 4-Dihydro-2, 6-dimethyl-3, 5-pyridinedicarboxylate on a Glassy Carbon Electrode.
Mechanistic Aspects of the Electrochemical Oxidation of Diethyl 1, 4-Dihydro-2, 6-dimethyl-3, 5-pyridinedicarboxylate on a Glassy Carbon Electrode.
Electroanalysis (1999)
Montoya MR, et al.