Merck
  • Controlling Selectivity in the Consecutive Reaction Network of Aldoxime Hydrogenation to Primary Amines.
Controlling Selectivity in the Consecutive Reaction Network of Aldoxime Hydrogenation to Primary Amines.
Synthesis
Gebauer-Henke E, et al., DOI: 10.1039/C2CY20356A