Merck
Effect of mechlorethamine hydrochloride (nitrogen mustard) on healing of abdominal wounds.
A.M.A. archives of surgery (1958-05-01)
S M FARHAT, N S AMER, D S WEEKS, M M MUSSELMAN
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%