Merck
  • Synthesis of azepino[4,5-b]indolones via an intermolecular radical oxidative substitution of N-Boc tryptamine.
Synthesis of azepino[4,5-b]indolones via an intermolecular radical oxidative substitution of N-Boc tryptamine.
Organic & biomolecular chemistry (2009-03-21)
Paul E Reyes-Gutiérrez, Rubén O Torres-Ochoa, Roberto Martínez, Luis D Miranda
ZUSAMMENFASSUNG
A two-step protocol for the synthesis of azepino[4,5-b]indolone derivatives featuring a xanthate radical oxidative aromatic substitution on the N-Boc protected tryptamine, using dilauroyl peroxide (DLP) as initiator and oxidant, is described.