Merck
  • A potential oncogenic role of the commonly observed E2F5 overexpression in hepatocellular carcinoma.
A potential oncogenic role of the commonly observed E2F5 overexpression in hepatocellular carcinoma.
World journal of gastroenterology (2011-01-29)
Yuzhu Jiang, Seon-Hee Yim, Hai-Dong Xu, Seung-Hyun Jung, So Young Yang, Hae-Jin Hu, Chan-Kwon Jung, Yeun-Jun Chung
ZUSAMMENFASSUNG
To explore the expression pattern of E2F5 in primary hepatocellular carcinomas (HCCs) and elucidate the roles of E2F5 in hepatocarcinogenesis. E2F5 expression was analyzed in 120 primary HCCs and 29 normal liver tissues by immunohistochemistry analysis. E2F5-small interfering RNA was transfected into HepG2, an E2F5-overexpressed HCC cell line. After E2F5 knockdown, cell growth capacity and migrating potential were examined. E2F5 was significantly overexpressed in primary HCCs compared with normal liver tissues (P = 0.008). The E2F5-silenced cells showed significantly reduced proliferation (P = 0.004). On the colony formation and soft agar assays, the number of colonies was significantly reduced in E2F5-silenced cells (P = 0.004 and P = 0.009, respectively). E2F5 knockdown resulted in the accumulation of G0/G1 phase cells and a reduction of S phase cells. The number of migrating/invading cells was also reduced after E2F5 knockdown (P = 0.021). To our knowledge, this is the first evidence that E2F5 is commonly overexpressed in primary HCC and that E2F5 knockdown significantly repressed the growth of HCC cells.