Merck
Aerobic biodegradation of amines in industrial saline wastewaters.
Chemosphere (2011-09-20)
Pablo Campo, William Platten, Makram T Suidan, Yunzhou Chai, John W Davis
ZUSAMMENFASSUNG
The treatment of hypersaline wastewaters represents a challenge since high salt concentrations disrupt bacteria present in normal biological treatments. This study was conducted to determine the fate of amines in two hypersaline wastewaters obtained from an industrial treatment plant processing influents with 3% and 7% of NaCl. The compounds were aniline (ANL), 4,4'-methylenedianiline (4,4'-MDA), cyclohexylamine (CHA), N-(2-aminoethyl)ethanolamine (AEA), N,N-diethylethanolamine (DEA), N,N-bis(2-hydroxyethyl)methylamine (MDEA), and tris(2-hydroxyethyl)amine (TEA). Mixtures of these chemicals with a mixed liquor suspended solids concentration of 1000 mg L(-1) were prepared at two salinities (3% and 7% NaCl). Ethanolamines were readily biodegraded at both salinities, following first-order kinetics with half-lives ranging between 10 and 58 h. Hydroxyl groups present in the ethanolamines had a positive impact on the biodegradation. Salinity did not affect the biodegradation rate of TEA and MDEA, whereas AEA and DEA degraded faster in 3% NaCl. After 48h, CHA was metabolized within a 24-h period in 3% NaCl, while no degradation was observed in 7% NaCl. ANL exhibited lag phases in both salinities and, in the following 24-h period, ANL concentrations dropped 40% and disappeared after 48 h. 4,4'-MDA degraded in 3% NaCl (half-life of 123 h) and remained unaltered after 120 h in 7% NaCl.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylendiamin, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.