Merck
  • HSpin1, a transmembrane protein interacting with Bcl-2/Bcl-xL, induces a caspase-independent autophagic cell death.
HSpin1, a transmembrane protein interacting with Bcl-2/Bcl-xL, induces a caspase-independent autophagic cell death.
Cell death and differentiation (2003-06-20)
H Yanagisawa, T Miyashita, Y Nakano, D Yamamoto
ZUSAMMENFASSUNG
The Drosophila spinster (spin) gene product is required for programmed cell death in the nervous and reproductive systems. We have identified a human homologue of the Drosophila spin gene product (HSpin1). HSpin1 bound to Bcl-2 and apoptosis regulator Bcl-X (Bcl-xL), but not to proapoptotic members such as Bcl-2-associated X protein and Bcl-2 homologous antagonist killer, in cells treated with TNF-alpha. Exogenous expression of HSpin1 resulted in the cell death without inducing a release of cytochrome c from mitochondria. Overexpression of Bcl-xL inhibited the HSpin1-induced cell death. Interestingly, a necrosis inhibitor, pyrrolidine dithiocarbomate, but not the pancaspase inhibitors, carbobenzoxy-VAD-fluoromethyl ketone and p35, blocked the HSpin1-induced cell death. HSpin1-induced cell death increases autophagic vacuole and mature form of cathepsin D, suggesting a novel caspase-independent cell death, which is link to autophagy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)